SY3H5815F433 Synthonix Corporation

1-tert-Butyl 4-methyl 1H-indole-1,4-dicarboxylate

  • Cas Number : 220499-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0914.html

Safety Information

MSDS https://synthonix.com/0914.html
Availability details for
Quantity
  • Availability
  • Add to Cart