SY3H5815F439 Synthonix Corporation

1-Benzylpyrrolidine-3-carbonitrile oxalate

  • Cas Number : 1188265-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/1268.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1268.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart