SY3H5815F441 Synthonix Corporation

2-Bromo-5-fluoroisonicotinaldehyde

  • Cas Number : 1005291-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/15938.html

Safety Information

MSDS https://synthonix.com/15938.html
Availability details for
Quantity
  • Availability
  • Add to Cart