SY3H5815F446 Synthonix Corporation

tert-Butyl 4-(4-amino-5-isopropoxy-2-methylphenyl)piperidine-1-carboxylate

  • Cas Number : 1032903-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16151.html

Safety Information

MSDS https://synthonix.com/16151.html
Availability details for
Quantity
  • Availability
  • Add to Cart