SY3H5815F45D Synthonix Corporation

(4-Benzylmorpholin-2-yl)methanamine

  • Cas Number : 110859-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16594.html

Safety Information

MSDS https://synthonix.com/16594.html
Availability details for
Quantity
  • Availability
  • Add to Cart