SY3H5815F460 Synthonix Corporation

5-Bromo-3-methoxy-2-methylpyridine

  • Cas Number : 1150617-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16799.html

Safety Information

MSDS https://synthonix.com/16799.html
Availability details for
Quantity
  • Availability
  • Add to Cart