SY3H5815F465 Synthonix Corporation

tert-Butyl (1-(2,5-difluorophenyl)-1-oxopent-4-yn-2-yl)carbamate

  • Cas Number : 1172623-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16948.html

Safety Information

MSDS https://synthonix.com/16948.html
Availability details for
Quantity
  • Availability
  • Add to Cart