SY3H5815F470 Synthonix Corporation

5-Bromo-6-fluoro-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 1207625-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17486.html

Safety Information

MSDS https://synthonix.com/17486.html
Availability details for
Quantity
  • Availability
  • Add to Cart