SY3H5815F473 Synthonix Corporation

2-Bromo-6-fluoro-3-methylpyridine

  • Cas Number : 1211536-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17536.html

Safety Information

MSDS https://synthonix.com/17536.html
Availability details for
Quantity
  • Availability
  • Add to Cart