SY3H5815F47A Synthonix Corporation

(6-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid

  • Cas Number : 1217500-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17616.html

Safety Information

MSDS https://synthonix.com/17616.html
Availability details for
Quantity
  • Availability
  • Add to Cart