SY3H5815F47E Synthonix Corporation

3-Bromo-2-methyl-6-nitropyridine

  • Cas Number : 1231930-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17800.html

Safety Information

MSDS https://synthonix.com/17800.html
Availability details for
Quantity
  • Availability
  • Add to Cart