SY3H5815F480 Synthonix Corporation

5-Bromo-4-methoxypyridin-2-amine

  • Cas Number : 1232431-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17813.html

Safety Information

MSDS https://synthonix.com/17813.html
Availability details for
Quantity
  • Availability
  • Add to Cart