SY3H5815F484 Synthonix Corporation

3-Bromo-1H-pyrazolo[3,4-d]pyrimidine

  • Cas Number : 1251033-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18019.html

Safety Information

MSDS https://synthonix.com/18019.html
Availability details for
Quantity
  • Availability
  • Add to Cart