SY3H5815F485 Synthonix Corporation

4-Bromo-2,3-dihydro-1H-inden-2-ol

  • Cas Number : 125141-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18025.html

Safety Information

MSDS https://synthonix.com/18025.html
Availability details for
Quantity
  • Availability
  • Add to Cart