SY3H5815F489 Synthonix Corporation

4-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 1256824-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18099.html

Safety Information

MSDS https://synthonix.com/18099.html
Availability details for
Quantity
  • Availability
  • Add to Cart