SY3H5815F48E Synthonix Corporation

5-Bromo-3-isopropyl-1H-indazole

  • Cas Number : 1276075-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18395.html

Safety Information

MSDS https://synthonix.com/18395.html
Availability details for
Quantity
  • Availability
  • Add to Cart