SY3H5815F491 Synthonix Corporation

4-Bromo-2-chloro-3-methylpyridine

  • Cas Number : 128071-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18413.html

Safety Information

MSDS https://synthonix.com/18413.html
Availability details for
Quantity
  • Availability
  • Add to Cart