SY3H5815F492 Synthonix Corporation

6-Benzyl-octahydropyrrolo[3,4-b]pyridine

  • Cas Number : 128740-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18450.html

Safety Information

MSDS https://synthonix.com/18450.html
Availability details for
Quantity
  • Availability
  • Add to Cart