SY3H5815F499 Synthonix Corporation

6-Bromo-1-isopropyl-1H-indazole-4-carboxylic acid

  • Cas Number : 1346702-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18826.html

Safety Information

MSDS https://synthonix.com/18826.html
Availability details for
Quantity
  • Availability
  • Add to Cart