SY3H5815F49A Synthonix Corporation

N-Boc-2-thiomorpholinecarboxylic Acid

  • Cas Number : 134676-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18827.html

Safety Information

MSDS https://synthonix.com/18827.html
Availability details for
Quantity
  • Availability
  • Add to Cart