SY3H5815F4A1 Synthonix Corporation

7-Bromo-5H-pyrrolo[3,2-d]pyrimidine

  • Cas Number : 1378816-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19223.html

Safety Information

MSDS https://synthonix.com/19223.html
Availability details for
Quantity
  • Availability
  • Add to Cart