SY3H5815F4A3 Synthonix Corporation

6-Bromo-5-fluoro-3-methyl-1H-indazole

  • Cas Number : 1394120-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19374.html

Safety Information

MSDS https://synthonix.com/19374.html
Availability details for
Quantity
  • Availability
  • Add to Cart