SY3H5815F4A5 Synthonix Corporation

4-Bromo-1,3-dihydroisobenzofuran

  • Cas Number : 1402667-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19438.html

Safety Information

MSDS https://synthonix.com/19438.html
Availability details for
Quantity
  • Availability
  • Add to Cart