SY3H5815F4B0 Synthonix Corporation

1-(tert-Butyl)-1H-pyrazole-4-carbonitrile

  • Cas Number : 149139-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20064.html

Safety Information

MSDS https://synthonix.com/20064.html
Availability details for
Quantity
  • Availability
  • Add to Cart