SY3H5815F4CF Synthonix Corporation

2-Bromo-4-ethoxypyridine

  • Cas Number : 17117-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21763.html

Safety Information

MSDS https://synthonix.com/21763.html
Availability details for
Quantity
  • Availability
  • Add to Cart