SY3H5815F4D4 Synthonix Corporation

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid

  • Cas Number : 175277-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21859.html

Safety Information

MSDS https://synthonix.com/21859.html
Availability details for
Quantity
  • Availability
  • Add to Cart