SY3H5815F505 Synthonix Corporation

5-Bromo-4-chloro-6-methylpyrimidine

  • Cas Number : 3438-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23168.html

Safety Information

MSDS https://synthonix.com/23168.html
Availability details for
Quantity
  • Availability
  • Add to Cart