SY3H5815F512 Synthonix Corporation

[2,3'-Bipyridin]-6'(1'H)-one

  • Cas Number : 381233-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23398.html

Safety Information

MSDS https://synthonix.com/23398.html
Availability details for
Quantity
  • Availability
  • Add to Cart