SY3H5815F515 Synthonix Corporation

2-Isobutyrylcyclohexanone

  • Cas Number : 39207-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23455.html

Safety Information

MSDS https://synthonix.com/23455.html
Availability details for
Quantity
  • Availability
  • Add to Cart