SY3H5815F51E Synthonix Corporation

1H-Benzo[d]imidazol-7-amine

  • Cas Number : 4331-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23667.html

Safety Information

MSDS https://synthonix.com/23667.html
Availability details for
Quantity
  • Availability
  • Add to Cart