SY3H5815F521 Synthonix Corporation

4-Bromo-N-methylpyridin-2-amine

  • Cas Number : 468718-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23788.html

Safety Information

MSDS https://synthonix.com/23788.html
Availability details for
Quantity
  • Availability
  • Add to Cart