SY3H5815F522 Synthonix Corporation

Benzofuran-3-carbaldehyde

  • Cas Number : 4687-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23789.html

Safety Information

MSDS https://synthonix.com/23789.html
Availability details for
Quantity
  • Availability
  • Add to Cart