SY3H5815F526 Synthonix Corporation

2-Bromo-4,6-dimethylpyridine

  • Cas Number : 4926-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23884.html

Safety Information

MSDS https://synthonix.com/23884.html
Availability details for
Quantity
  • Availability
  • Add to Cart