SY3H5815F531 Synthonix Corporation

1-(2-Bromo-6-hydroxyphenyl)ethanone

  • Cas Number : 55736-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24263.html

Safety Information

MSDS https://synthonix.com/24263.html
Availability details for
Quantity
  • Availability
  • Add to Cart