SY3H5815F537 Synthonix Corporation

Benzo[b]thiophen-6-ylmethanol

  • Cas Number : 6179-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24567.html

Safety Information

MSDS https://synthonix.com/24567.html
Availability details for
Quantity
  • Availability
  • Add to Cart