SY3H5815F544 Synthonix Corporation

5-Bromo-8-methoxyisoquinoline

  • Cas Number : 679433-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24866.html

Safety Information

MSDS https://synthonix.com/24866.html
Availability details for
Quantity
  • Availability
  • Add to Cart