SY3H5815F54A Synthonix Corporation

Benzo[d][1,3]dioxol-4-ol

  • Cas Number : 69393-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24932.html

Safety Information

MSDS https://synthonix.com/24932.html
Availability details for
Quantity
  • Availability
  • Add to Cart