SY3H5815F553 Synthonix Corporation

tert-Butyl 3-vinylpyrrolidine-1-carboxylate

  • Cas Number : 753015-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25220.html

Safety Information

MSDS https://synthonix.com/25220.html
Availability details for
Quantity
  • Availability
  • Add to Cart