SY3H5815F556 Synthonix Corporation

6-Bromo-2,3-dihydroquinolin-4(1H)-one

  • Cas Number : 76228-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25273.html

Safety Information

MSDS https://synthonix.com/25273.html
Availability details for
Quantity
  • Availability
  • Add to Cart