SY3H5815F56D Synthonix Corporation

5-Bromo-4-chloro-2-methylpyrimidine

  • Cas Number : 861383-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25786.html

Safety Information

MSDS https://synthonix.com/25786.html
Availability details for
Quantity
  • Availability
  • Add to Cart