SY3H5815F56F Synthonix Corporation

1-(5-Bromo-2,4-difluorophenyl)ethanone

  • Cas Number : 864773-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25842.html

Safety Information

MSDS https://synthonix.com/25842.html
Availability details for
Quantity
  • Availability
  • Add to Cart