SY3H5815F57E Synthonix Corporation

3-Bromo-1H-indazol-4-amine

  • Cas Number : 885521-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26148.html

Safety Information

MSDS https://synthonix.com/26148.html
Availability details for
Quantity
  • Availability
  • Add to Cart