SY3H5815F582 Synthonix Corporation

tert-Butyl 3-(cyclopropylamino)pyrrolidine-1-carboxylate

  • Cas Number : 887587-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26205.html

Safety Information

MSDS https://synthonix.com/26205.html
Availability details for
Quantity
  • Availability
  • Add to Cart