SY3H5815F58A Synthonix Corporation

2,8-Bis(trifluoromethyl)-4-hydroxyquinoline

  • Cas Number : 35853-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2643.html

Safety Information

MSDS https://synthonix.com/2643.html
Availability details for
Quantity
  • Availability
  • Add to Cart