SY3H5815F58C Synthonix Corporation

4-((Benzyloxy)methyl)-6-chloropyrimidine

  • Cas Number : 914802-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26443.html

Safety Information

MSDS https://synthonix.com/26443.html
Availability details for
Quantity
  • Availability
  • Add to Cart