SY3H5815F58E Synthonix Corporation

4-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 916176-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26471.html

Safety Information

MSDS https://synthonix.com/26471.html
Availability details for
Quantity
  • Availability
  • Add to Cart