SY3H5815F591 Synthonix Corporation

5-Bromo-3-methyl-1H-pyrazolo[3,4-c]pyridine

  • Cas Number : 929617-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26576.html

Safety Information

MSDS https://synthonix.com/26576.html
Availability details for
Quantity
  • Availability
  • Add to Cart