SY3H5815F599 Synthonix Corporation

(R)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate hydrochloride

  • Cas Number : 947275-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26803.html

Safety Information

MSDS https://synthonix.com/26803.html
Availability details for
Quantity
  • Availability
  • Add to Cart