SY3H5815F59B Synthonix Corporation

2-Bromo-5-fluoro-4-nitroaniline

  • Cas Number : 952664-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26858.html

Safety Information

MSDS https://synthonix.com/26858.html
Availability details for
Quantity
  • Availability
  • Add to Cart