SY3H5815F5BF Synthonix Corporation

4-(Benzyloxy)-2-methoxybenzaldehyde

  • Cas Number : 58026-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3195.html

Safety Information

MSDS https://synthonix.com/3195.html
Availability details for
Quantity
  • Availability
  • Add to Cart